Learn More
Corning™ Lymphocyte Serum-free Medium, KBM 581
The use of T-lymphocytes in adaptive immunotherapy is emerging as an important tool in the treatment of cancer and other clinical applications.
Supplier:  Corning™ 88581CM
Catalog No. MT88581CM
Only null left
Description
Description
Corning™'s Lymphocyte Serum Free Medium (88-581-CM) supports the proliferation and activation of lymphocytes, including tumor infiltrating lymphocytes (TIL), Cytokine-Induced Killer (CIK) cells and Natural Killer (NK) cells. There is technical data to support the use in CAR-T as well as several other important immunotherapy applications. The 88-581-CM medium is specialized for both activation and expansion of human CIK cells. This enhanced version contains more nutrients, shows improved proliferation capacity, and produces a higher ratio of CIK cells (CD3+ CD56+).
Specifications
Specifications
Liquid
1000 mL
Product Suggestions
Product Suggestions