missing translation for 'onlineSavingsMsg'
Learn More

Lycopene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. p-7187455
Change view
Click to view available options
Quantity:
500 mg
5 x 100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014357 5 x 100 mg
111014358 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014357 Supplier Merck Emd Millipore Supplier No. PHR17705X100MG
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 502-65-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C40H56
MDL Number MFCD00017350
Synonym Lycopene; psi,psi-Carotene; 2,6,10,14,19,23,27,31-Octamethyl-dotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
Type Certified Reference Material
Quantity 5 x 100 mg
Formula Weight 536.87
Grade Certified Reference Material