missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ly-49H Monoclonal Antibody (3D10), PE, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 12588682

Encompass

 View more versions of this product

Catalog No. 12588682

Only null left

Description

Description

Description: The monoclonal antibody 3D10 recognizes Ly49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly49H, like Ly49D, lacks the characteristic ITAM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly49H is expressed in a subset of NK cells but not NKT (NK1.1/CD3+) cells. Natural killer cells can express several Ly49 proteins. It has been shown that there is preferential staining of Ly49H with Ly49D(Product # 12-5783-81) . Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of monoclonal antibody 3D10 to cell cultures induces lysis of target cells. Applications Reported:This 3D10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3D10 antibody has been tested by flow cytometric analyis of C57Bl/6 splenocytes. This can be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Each NovaFluor conjugate or kit is shipped with CellBlox Blockin...

Ly49H is a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly49H, like Ly49D, lacks the characteristic ITAM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly49H is expressed in a subset of NK cells but not NKT (NK1.1/CD3+) cells. Natural killer cells can express several Ly49 proteins.
Specifications

Chemical Identifiers

Ly-49H
0.2 mg/mL
Flow Cytometry
PE
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
Q60682
16639
Primary
Affinity chromatography
3D10
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG1 κ
Mouse
Klra8
Cmv1; Cmv-1; cytomegalovirus resistance 1; killer cell lectin-like receptor 8; killer cell lectin-like receptor, subfamily A, member 8; Klra8; Ly49H; Ly-49h; Ly49-h; Ly49u>129>; Lymphocyte antigen 49h; T-cell surface glycoprotein Ly-49H
Klra8
Antibody

Specifications

Ly-49H
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q60682
Klra8
Affinity chromatography
RUO
16639
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
3D10
PE
Klra8
Cmv1; Cmv-1; cytomegalovirus resistance 1; killer cell lectin-like receptor 8; killer cell lectin-like receptor, subfamily A, member 8; Klra8; Ly49H; Ly-49h; Ly49-h; Ly49u>129>; Lymphocyte antigen 49h; T-cell surface glycoprotein Ly-49H
Mouse
100 μg
Primary
Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions