missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ly-49G Monoclonal Antibody (AT-8 (AT8)), PE, eBioscience™

Catalog No. 5010948
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5010948 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5010948 Supplier Invitrogen™ Supplier No. 12588582

Mouse Monoclonal Antibody

Description: The AT8 monoclonal antibody reacts with the mouse Ly-49G receptor from 129, FVB, BALB/c, C3H, CBA, SJL and A/J, but not C57Bl/6 mice. Ly49G, a member of the Ly49 family is expressed by NK cell subset. Applications Reported: The AT-8 (AT8) antibody has been reported for use in flow cytometric analysis. Applications Tested: The AT-8 (AT8) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.

Specifications

Antigen Ly-49G
Applications Flow Cytometry
Classification Monoclonal
Clone AT-8 (AT8)
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra7
Gene Accession No. Q60654
Gene Alias Killer cell lectin-like receptor 7; killer cell lectin-like receptor, subfamily A, member 7; Klra7; LGL-1; Ly49g; ly-49g; Ly49-g; Ly-49G.1; Ly-49G.2; Ly-49G.3; Ly49G.4; Ly49g1; Ly49g2; Ly49g4; Lymphocyte antigen 49g; T-cell surface glycoprotein Ly-49G
Gene Symbols Klra7
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16638
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less