Learn More
Applied Biosystems™ LV200™ dT Pack
Description
Save Time and MoneyCompared to conventional columns, the ABI LV200™ columns reduce cycle time by about 10% and the cost of synthesis by about 40%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments
- Proprietary design and manufacturing methods ensure consistency and high quality
- Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
- Hydrophobic polystyrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis
- The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length
Specifications
Specifications
| Packaging Type | Shipped at Room Temperature |
| Quantity | 60 Columns |
For Research Use Only. Not for use in diagnostic procedures.