Learn More
Applied Biosystems™ LV200™ dG(iBu) Pack
Catalog No. 4316442
Encompass
Change view
Click to view available options
Quantity:
60 Columns
Catalog No. Quantity
4316442 60 Columns
1 options
Catalog No. 4316442 Supplier Applied Biosystems™ Supplier No. 4316442
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Contains 60 dG(iBu) ABI LV200™ synthesis columns.The dG(iBu) ABI LV200™ synthesis columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.
Save Time and MoneyCompared to conventional columns, the ABI LV200™ columns reduce cycle time by about 10% and the cost of synthesis by about 40%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
  • Proprietary design and manufacturing methods ensure consistency and high quality
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
  • Hydrophobic polystyrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis
  • The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length
Specifications
Packaging Type Shipped at Room Temperature
Quantity 60 Columns
For Research Use Only. Not for use in diagnostic procedures.