missing translation for 'onlineSavingsMsg'
Learn More

CTS™ CTS™ LV-MAX™ Transfection Kit

Catalog No. A4132601
Encompass
Change view
Click to view available options
Quantity:
1 Kit (For 1 L of LV Production)
1 Kit (For 10 L of LV Production)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4132601 1 Kit (For 1 L of LV Production)
A4132602 1 Kit (For 10 L of LV Production)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4132601 Supplier CTS™ Supplier No. A4132601

The Gibco CTS LV-MAX Transfection Kit is an essential component of the CTS LV-MAX lentiviral production system.

The Gibco CTS LV-MAX Transfection Kit is an essential component of the CTS LV-MAX lentiviral production system. Based on extensive Design of Experiment (DOE) methodologies, the cationic lipid-based transfection reagent, lentiviral production supplement, and LV enhancer are specifically screened for maximal delivery of multiple DNA plasmids to achieve the highest possible lentiviral titer when used in conjunction with CTS Viral Production Cells cultured in CTS LV-MAX Production Medium.

All CTS LV-MAX Transfection Kit components are produced under cGMP manufacturing conditions with extensive quality-control testing to ensure proper preclinical, clinical, and commercial therapeutic viral vector applications.

CTS LV-MAX Transfection Kit features include:

  • Uniquely designed to enhance transient transfection of multiple DNA plasmids into high density suspension HEK293-derived cell culture
  • Enables high viral titers of up to 1 x 108 TU/mL (unconcentrated LVV-GFP) to allow for a cost-effective alternative to PEI-based transfection reagents
  • Scalable protocol for 96-deep well blocks, flasks, and bioreactors
  • Regulatory-compliant reagents produced in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.

The CTS LV-MAX Transfection Kit requires CTS Opti-MEM I (not included in this kit) for complexation. The chemically-defined, serum-free, animal origin-free features of the CTS LV-MAX Transfection Kit result in high reproducibility and lot-to-lot reliability. The formulation of this product is identical to the research-grade LV-MAX Transfection Kit, but the CTS LV-MAX Transfection Kit is validated for clinical and commercial therapeutic applications.

Custom configuration and packaging available upon request.

Specifications

Classification Animal Origin-Free
Content And Storage • CTS LV-MAX Supplement, 50 mL
• CTS LV-MAX Transfection Reagent, 6 mL
• CTS LV-MAX Enhancer, 40 mL

Store at 2–8°C and protect from light. Ships on gel ice.

Culture Type Mammalian
Cell Type HEK293
Format Bottle
For Use With (Application) LV Production
Product Type Transfection Kit
Quantity 1 Kit (For 1 L of LV Production)
Reagent Type Cationic Lipid-Based Transfection
Shipping Condition Gel Ice
Manufacturing Quality ISO 13485, FDA-registered, 21 CFR 820
Show More Show Less
Why do you recommend using V bottom 96-well plates for harvesting lentivirus from CTS Viral Production Cells?

We recommend using V-bottom 96-well plates to harvest the supernatant containing lentiviral particles as they allow for easier detection of the cell pellets after centrifugation of the cultures.

During lentivirus production from CTS Viral Production Cells, will the shaker vibration affect the cell growth of other cultures maintained in the same incubator or in stacked incubators?

We recommend maintaining shaking cultures in a separate incubator from all other incubators to avoid vibration effects. Alternatively, we recommend using a small, quiet shaker such as the Thermo Scientific CO2 Resistant Shaker (Cat. No. 88881102) for convenience and best performance.

Is 8% CO2 a growth requirement for CTS Viral Production Cells?

We generally recommend culturing the CTS Viral Production Cells under 8% CO2 conditions. We found that cells cultured in 5% CO2 grew slower and were more prone to more cell clumping than when grown in 8% CO2.

What is special about the CTS LV-MAX lentivirus production system?

The CTS LV-MAX lentivirus production system is a fully-optimized complete system in which the cells, media, and transfection reagent work together to generate high-titer lentivirus. The CTS LV-MAX Production Medium is specifically designed to work with the system, to support high density suspension cell growth of CTS Viral Production Cells.

The manual recommends using CTS Viral Production Cells passaged to P5 (˜3 weeks) on the day of transfection. Are these general cell culture guidelines or special requirements for these cells?

Although 3-4 passages before transfection are sufficient for most cell lines recovering directly from liquid N2 storage, we found that CTS Virus Production Cells passaged to P5 showed the highest performance in lentivirus particle production and reproducibility between batches. Therefore, we recommend passaging the cells to P4 at the end of a given week for lentivirus production the following week.

The manual recommends passaging the CTS Viral Production Cells three times before freezing. Can I bank the cells sooner than that?

For optimal cell recovery from liquid N2 storage, CTS Viral Production Cells should be passaged to passage 2 (P2) (from the original vial) and up to P3 before banking, to ensure good cell health and viability. Cells that demonstrate good cell health (i.e., ˜26 hr doubling time, round cell morphology, and minimal clumping) may be banked at P2 or P3.

The CTS LV-MAX lentivirus production system user manual recommends the passage of CTS Viral Production Cells every 3-4 days; is this necessary? Can I seed the cells less densely and passage less frequently (i.e., every 6-7 days)?

Passaging the CTS Viral Production Cells every 3-4 days allows for cell growth rates to progress from lag to log phase, thus ensuring highest cell viability after splitting cultures. We recommend passaging the cells at a density of no lower than ˜0.35x10E6 viable cells/mL as low cell densities maintained in the lag phase for long periods, will eventually lead to poor overall cell growth. We do not recommend culturing the cells past 4 days.
The cell culture densities for CTS Viral Production Cells are as follows:
- 0.35x10E6 and 0.55x10E6 viable cells/mL (splitting cell density for 4 day and 3 day culture, respectively)
- 3.5x10E6 to 5x10E6 viable cells/mL (subculture cell density)
- Log-phase is from 0.35x10E6 to 6.5x10E6 viable cells/mL.

With the CTS LV-MAX lentivirus production system, do you have any recommendations for how I can produce more lentiviral expression plasmid?

When propagating lentiviral expression plasmids, it is important to propagate them in cells that will minimize any sequence rearrangements or unwanted recombination due to the long terminal repeat (LTRs) sequences and large size of the vector backbone ( ˜10 Kb). To minimize these events, we recommend using Stbl3 cells for stable propagation of lentiviral expression plasmids.

With the CTS LV-MAX lentivirus production system, I would like to titer my lentivirus using a different method from GFP or antibiotic method, is this okay?

We recommend using either the GFP method or antibiotic-based method described in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf). You may try other viral titering methods, however, we do not have protocols for methods other than by GFP or antibiotics.

I am using the CTS LV-MAX lentivirus production system and my lentiviral titer is low. How do I troubleshoot effectively and what can I do to optimize this?

Low lentiviral titer could be due to many different factors such as non-optimized lentiviral expression vector design, low transfection efficiency, incorrect cell density, suboptimal culture conditions (e.g., incubator, shaker speed, vessel type, media contamination), purification strategy, and titer method. To best track your workflow from culturing your cells to titering the lentivirus, we recommend using a positive control such as the Vivid Colors pLenti6.3/V5-GW/EmGFP Expression Control Vector (Cat. No. V37006).

I am using CTS Viral Production Cells and am seeing many dead cells in my culture. What could be causing this?

This could mean that the seeding or starting cell density was too low. The CTS LV-MAX Supplement relies on high cell density. Refer to the CTS LV-MAX Lentiviral Production System manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf) for cell density recommendations.

My CTS Viral Production Cells are clumping. Can I use an anti-clumping reagent?

Anti-clumping reagents could potentially inhibit transfection efficiency and are generally not recommended. If needed, add no more than 0.1% Pluronic F-68

Can I use other plasmids with the CTS LV-MAX lentivirus production system?

The CTS LV-MAX lentivirus production system is designed to be compatible with any format of plasmid DNA. The quality of plasmids (e.g., purity), ratio of the plasmids, and vector design all contribute to lentiviral titer. Please contact us at techsupport@thermofisher.com for suggestions.

In the CTS LV-MAX transfection protocol, CTS LV-MAX Enhancer addition time is suggested to be anytime between 5-14 hr after transfection. How critical is it that I stay within this time window?

The formulation of the CTS LV-MAX Enhancer coupled with the broad window of enhancer addition time was specifically optimized to allow user flexibility in schedule and start time of the protocol without affecting performance. For optimal results, we recommend adhering to the suggested time window for enhancer addition.

In the CTS LV-MAX protocol, the cells remain in the same media throughout the two-day transfection. Why is no media change needed?

The CTS LV-MAX Production Medium is specially designed to support the health of Viral Production Cells cultured at high-density. We have performed comprehensive studies in-house and have determined that there is no need for media change during the protocol.

Do you have recommendations for measuring CTS lentiviral titer using a non-fluorescent method?

Please refer to page 31 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf) for a protocol for lentiviral titer measurement using antibiotic selection.

Are the sequences of the LV-MAX Lentiviral Packaging Mix plasmids confidential?

The sequences of the LV-MAX Lentiviral Packaging Mix plasmids are published on our website; however, the ratio of the plasmids in the mix is confidential.

What is the recommended ratio of plasmids to use in the CTS LV-MAX lentivirus production system?

We recommend using the lentiviral packaging plasmids and transfer plasmids at a ratio of 3:2.

Should I harvest the lentivirus from the CTS Viral Production Cells at 24 hr and 48 hr time-points. How about waiting for 72 hr?

Our system has been optimized to provide maximal lentiviral production at 48 hr post transfection. Waiting until 72 hr is not recommended since existing lentiviral particles within the media will infect the production cells and can reduce the titer.

The CTS Viral Production Cells are supposed to be good in passage 5-20. Does this refer to passage 5-20 post-thaw, or should we consider the number of passages the cells went through before cryopreservation?

The passages should be counted upon thawing, and the cells should be discarded when they reach passage 20 post-thaw.

Can I transfect CTS Viral Production Cells before passage 5 (P5)?

We recommend that CTS Viral Production Cells be used for lentiviral production only following passage 5 to allow for full recovery of the cells from the cryopreservation; CTS Viral Production Cells are sensitive upon thawing and need the extra passages for recovery.

Why do I need to dilute the CTS Viral Production Cells once more before transfection?

This is a necessary step to replenish nutrients and ensure that the cells are at the correct density to achieve the concentration needed for optimal transfection efficiency. It is crucial that the cells are at the proper cell density during the time of transfection to ensure maximal performance.

What density should I be seeding the CTS Viral Production Cells at?

Depending on whether your culture will be 3 or 4 days, we have outlined the differences in seeding densities in the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf). We also have specific guidance for seeding densities to be used for lentiviral vector production.

Do you have protocols you can share for scale-up with the CTS LV-MAX lentivirus production system? What scale have you tested so far?

We continually develop large-scale protocols to support clinical and commercial lentiviral vector production. Currently, we have a 2 L bio-reactor protocol that is available from techsupport@thermofisher.com. We are also working on a 5 L and 50 L scale.

My CTS Viral Production Cells have low viability at 3 days post-thawing. Is this normal?

It is okay to see variation in viability upon thawing, but this should normalize during subsequent passages. We recommend culturing the cells as described in chapter 2 of the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018450_CTS_LV-MAX_LentiviralProdSys_UG.pdf) so that the cells recover sufficiently and achieve >90% viability before starting the protocol for CTS lentviral production.

Can I use other media like HEK293 expression media with the CTS LV-MAX products?

We have tried other media and have seen reduced performance. For maximal lentiviral vector yield, we recommend using the CTS LV-MAX Production Medium.

Can I use other commercially available transfection reagents with the CTS LV-MAX Production Medium and CTS Viral Production Cells?

The CTS LV-MAX Transfection Kit is specifically designed to work synergistically with the CTS LV-MAX Production Medium and CTS Viral Production Cells to provide maximal lentiviral titer. Mixing and matching reagents is not recommended, as this will decrease the overall performance.

I'm interested only in the CTS LV-MAX Transfection Reagent. Can I purchase it separately from the CTS LV-MAX Transfection Kit?

The CTS LV-MAX Transfection Kit contains three components, the transfection reagent, supplement, and enhancer, that are not sold as stand-alone items. The system components were specifically optimized to work as a complete system to obtain maximal lentiviral titer. We can however customize different ratios of the transfection reagent, supplement, and enhancer - for further information, please send an email to customorders@thermofisher.com.

Do you have a protocol to detect residual components of the CTS LV-MAX transfection reagent and enhancer?

We can provide protocol guidance for how to detect transfection reagent components under CDA. Please contact our outlicensing@thermofisher.com for information.

Do you have any guidelines for purification of lentivirus from CTS Viral Production Cells under serum-free conditions?

For downstream purification of lentivirus under serum-free conditions, we recommend using tangential flow filtration. Please contact techsupport@thermofisher.com for the suggested protocol. In order to maintain the integrity of lentiviral vectors, we do not recommend using ultracentrifugation under serum-free conditions.

What is the difference between Expi293F Cells, Viral Production Cells, CTS Viral Production Cells, and FreeStyle 293-F Cells?

The intended use, validation, and protocol optimization varies among the different cell types. Expi293F Cells and FreeStyle 293-F Cells were specifically developed and validated for transient protein production while Viral Production Cells and CTS Viral Production Cells were designed and validated for lentiviral vector manufacturing. The commonality between the Expi293F Cells, Viral Production Cells, and CTS Viral Production Cells is that they were originally derived from FreeStyle 293-F lineage. Both Expi293F and Freestyle 293F cells come with regulatory support intended for bio-production applications whereas CTS Viral Production Cells (cGMP banked) are intended for cell and gene therapy applications, and complemented with a more robust regulatory support package to enable clinical gene therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.

What are the major differences between LV-MAX and CTS LV- MAX products?

The LV-MAX and CTS LV- MAX products have equivalent performance. In contrast to the research use only LV-MAX products, CTS LV-MAX products have an additional intended use for cell and gene therapy applications, and are complemented with a more robust regulatory support package to enable clinical therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.

What is the intended use claim for your LV-MAX and CTS LV-MAX products?

Our LV-MAX products are intended for research use only while our CTS LV- MAX products are intended for research use or manufacture of cell, gene or tissue based products. Please refer to the individual products for further details on the intended use claim.

What kind of documentation do you provide with the CTS Viral Production Cells?

We have extensive documentation to demonstrate lineage history, viral clearance, and traceability of our GMP manufactured process. Under CDA, we can share our Certificate of Analysis and Table of Contents. Full access to the documentation package is available through outlicensing@thermofisher.com.

Can I use the CTS LV-MAX products for clinical trials or commercial use?

Please refer to our standard sales terms and conditions and limited use label license. For further inquiries, please send an email to outlicensing@thermofisher.com.


For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.