missing translation for 'onlineSavingsMsg'
Learn More

Betaine, Anhydrous, Spectrum™ Chemical

Catalog No. 18609067
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609067 500 g Poly Bottle
18609064 100 g Poly Bottle
18609065 2.5 kg Poly Bottle
18609066 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609067 Supplier Spectrum Chemical Mfg Cor Supplier No. B1572500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-43-7

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Molecular Weight (g/mol) 117.15
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material Betaine, Anhydrous

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.