missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) oxide, REacton™, 99.99% (REO)

Catalog No. p-4781978
missing translation for 'changeView'
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1125603 1 g
AA1125606 5 g
AA1125614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AA1125603 Supplier Thermo Scientific Chemicals Supplier No. 01125603
Only null left

CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol

Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Melting Point 2487°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water.
Formula Weight 397.94
Odor Odorless
Sensitivity Hygroscopic
Density 9.420 g/cm3 at 20°C
Chemical Name or Material Lutetium(III) oxide, Rare Earth Oxides Content
Show More Show Less
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-764-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only