Learn More
Lutetium(III) oxide, REacton™, 99.99% (REO)
Description
Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2487°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.99% (REO) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 397.94 |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 9.42 |
| Show More |
Safety and Handling
| Hazard Category | H315-H320-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 234-764-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only