missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luteolin 7-glucoside, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 5373-11-5 |
| Physical Form | Neat |
| Linear Formula | C21H20O11 |
| MDL Number | MFCD06799436 |
| Synonym | Luteolin 7-O-beta-D-glucoside; 3 ′,4 ′,5,7-Tetrahydroxyflavone 7-glucoside; Cynaroside; Glucoluteolin; Glucosylluteolin; Luteoloside |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 448.38 |
| Percent Purity | ≥98% (HPLC) |
| Show More |