missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lutein Solution, 1 mg/L in Ethanol, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 127-40-2 |
| Linear Formula | C40H56O2 |
| MDL Number | MFCD08435941 |
| Synonym | beta,epsilon-Carotene-3,3 ′-diol; Xanthophyll solution |
| Type | Analytical Standard |
| Quantity | 2.5 mL |
| Formula Weight | 568.87 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1 mg/L in ethanol |