missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Luminata Crescendo ELISA HRP Substrate

Catalog No. p-7078314
Change view
Click to view available options
Quantity:
100 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ELLUR0100 100 mL
ELLUR0200 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ELLUR0100 Supplier MilliporeSigma™ Supplier No. ELLUR0100
Only null left

Used on immunoblots as chemiluminescent substrate

  • Detection Methods: Chemiluminescence
  • Key Applications: ELISA
  • Chemiluminescent: HRP detection reagent
  • For use on immunoblots

Specifications

Quantity 100 mL
Content And Storage 2°C to 8°C