missing translation for 'onlineSavingsMsg'
Learn More

Luff-Schoorl Reagent, Honeywell Fluka™

Volumetric, 25 g/L

$192.89 - $789.39

Specifications

Chemical Name or Material Luff-Schoorl Reagent
CAS 497-19-8,7732-18-5,7758-98-7
UN Number UN3082
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000912
View Documents
Honeywell Chemicals
344001L
1 L
Each for $192.89
Only null left
 
6000913
View Documents
Honeywell-Fluka
344005LVP
5 L
Each for $789.39
Only null left
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Specifications

Luff-Schoorl Reagent
497-19-8,7732-18-5,7758-98-7
UN3082
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'