missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Luciferin Detection Reagent with Esterase
A reagent to detect presence of pro-luciferin substrates used to monitor the activity of specific isoforms of cytochrome P450 or NAT2.
$289.87 - $697.01
Specifications
| Content And Storage | -30°C to -10°C |
|---|---|
| For Use With (Application) | Monitor the activity of specific isoforms of cytochrome P450 or NAT2 |
| pH | 7.2 |
| Product Type | Luciferin |
Description
- Reagent for Detection of Pro-Luciferin Substrates
- Reagent available with esterase
- Use in combination with the pro-luciferin substrates
Specifications
| -30°C to -10°C | |
| Monitor the activity of specific isoforms of cytochrome P450 or NAT2 | |
| 7.2 | |
| Luciferin |