Learn More
Promega Luciferin Detection Reagent with Esterase
A reagent to detect presence of pro-luciferin substrates used to monitor the activity of specific isoforms of cytochrome P450 or NAT2.
Supplier:  Promega V8930
Encompass
 View more versions of this product
Catalog No. PRV8930
Only null left
Description
Description
  • Reagent for Detection of Pro-Luciferin Substrates
  • Reagent available with esterase
  • Use in combination with the pro-luciferin substrates
Specifications
Chemical Identifiers
-30°C to -10°C
Monitor the activity of specific isoforms of cytochrome P450 or NAT2
7.2
Luciferin
Specifications
10 mL
Monitor the activity of specific isoforms of cytochrome P450 or NAT2
7.2
-30°C to -10°C
10mL luciferin detection reagent and 10mL reconstitution buffer with esterase
Luciferin
Product Suggestions
Product Suggestions