missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LTK (TYK1), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic
LTK (TYK1), ALK and Ros have been identified as receptor tyrosine kinases having sequence similarity to the Insulin receptor subfamily of kinases. LTK, leukocyte tyrosine kinase, is expressed in murine B-lymphocyte precursors and has also been found in forebrain neurons. ALK, anaplastic lymphoma kinase, is normally highly expressed specifically in the nervous system. A truncated form containing the catalytic domain of ALK is expressed as the result of a translocation occurring in many non-Hodgkin's lymphomas. The c-Ros gene was originally identified in mutant form as an oncogene. Ros is normally expressed in a small number of epithelial cell types and may play a role in epithelial development.
Specifications
Specifications
| Accession Number | P29376 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 4058 |
| Molecular Weight (g/mol) | 73.7 kDa |
| Name | Human LTK (TYK1), GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | EC 2.7.1.112; EC 2.7.10.1; leukocyte receptor tyrosine kinase; leukocyte tyrosine kinase; leukocyte tyrosine kinase receptor; Ltk; Protein tyrosine kinase 1; TYK1; tyrosine kinase-1 |
| Show More |