missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5106479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106479 Supplier Invitrogen™ Supplier No. PA5106479
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LTK.

LTK (TYK1), ALK and Ros have been identified as receptor tyrosine kinases having sequence similarity to the Insulin receptor subfamily of kinases. LTK, leukocyte tyrosine kinase, is expressed in murine B-lymphocyte precursors and has also been found in forebrain neurons. ALK, anaplastic lymphoma kinase, is normally highly expressed specifically in the nervous system. A truncated form containing the catalytic domain of ALK is expressed as the result of a translocation occurring in many non-Hodgkin's lymphomas. The c-Ros gene was originally identified in mutant form as an oncogene. Ros is normally expressed in a small number of epithelial cell types and may play a role in epithelial development.

Specifications

Antigen LTK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LTK
Gene Accession No. P08923, P29376
Gene Alias EC 2.7.1.112; EC 2.7.10.1; leukocyte receptor tyrosine kinase; leukocyte tyrosine kinase; leukocyte tyrosine kinase receptor; Ltk; Protein tyrosine kinase 1; TYK1; tyrosine kinase-1
Gene Symbols LTK
Host Species Rabbit
Immunogen A synthesized peptide derived from human LTK(Accession P29376), corresponding to amino acid residues R221-A271.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17005, 311337, 4058
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less