missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRN4 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon (100%) Monkey (94%).
LRRN4 may play an important role in hippocampus-dependent long-lasting memory.
Specifications
Specifications
| Antigen | LRRN4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | LRRN4 |
| Gene Accession No. | Q8WUT4 |
| Gene Alias | C20orf75; dJ1056H1.1; leucine rich repeat neuronal 4; leucine-rich repeat neuronal protein 4; LRRN4; Neuronal leucine-rich repeat protein 4; NLRR4; NLRR-4 |
| Gene Symbols | LRRN4 |
| Show More |