missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRK2 Recombinant Rabbit Monoclonal Antibody (HL3167)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA566193
Description
LRRK2 Recombinant Monoclonal Antibody for Western Blot, IHC (F)
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Mutations in LRRK2 have been associated with Parkinson's disease. LRRK2 is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane.Chemical Identifiers
| LRRK2 | |
| 1 mg/mL | |
| Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Q5S006, Q5S007 | |
| 120892, 66725 | |
| The immunogen used to generate this antibody corresponds to human LRRK2. | |
| Antibody |
| HL3167 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| LRRK2 | |
| 4921513O20Rik; 9330188B09Rik; augmented in rheumatoid arthritis 17; AURA17; AW561911; BC 300-268; cI-46; D630001M17Rik; DARDARIN; DKFZp434H2111; FLJ45829; Gm927; leucine rich repeat kinase 2; leucine-rich repeat kinase 2; leucine-rich repeat serine/threonine-protein kinase 2; LRR kinase 2; LRRK 2; Lrrk2; LRRK-2; PARK8; RIPK7; ROCO2 | |
| LRRK2 | |
| Primary | |
| Protein A |
Specifications
| LRRK2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q5S006, Q5S007 | |
| LRRK2 | |
| The immunogen used to generate this antibody corresponds to human LRRK2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot | |
| HL3167 | |
| Unconjugated | |
| LRRK2 | |
| 4921513O20Rik; 9330188B09Rik; augmented in rheumatoid arthritis 17; AURA17; AW561911; BC 300-268; cI-46; D630001M17Rik; DARDARIN; DKFZp434H2111; FLJ45829; Gm927; leucine rich repeat kinase 2; leucine-rich repeat kinase 2; leucine-rich repeat serine/threonine-protein kinase 2; LRR kinase 2; LRRK 2; Lrrk2; LRRK-2; PARK8; RIPK7; ROCO2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 120892, 66725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |