missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRP8 Recombinant Rabbit Monoclonal Antibody (23GB2520)

Catalog No. MA553503
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553503 20 μL
MA553502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553503 Supplier Invitrogen™ Supplier No. MA553503
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

LRP8 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.

Specifications

Antigen LRP8
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2520
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LRP8
Gene Accession No. Q14114, Q924X6
Gene Alias 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B
Gene Symbols LRP8
Host Species Rabbit
Immunogen A synthesized peptide derived from human ApoER2 (900-963AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16975, 362558, 7804
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less