missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lrmp Monoclonal Antibody (K52026_16E2), Invitrogen™

Catalog No. MA556348
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556348 200 μL
MA555968 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556348 Supplier Invitrogen™ Supplier No. MA556348
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: LSGCIRMNDD PSMEENGVER VCPESLLQSR EYSSLPLPRH TSSTDGTITS SDPGLEILNM ASCDLDRNSL CKKEEDTRSA SPTIEAQGTS PAHDNIAFQD STSKDKTILN LEAKEEPETI EEHKKEHASG DSVVSPLPVT TVKSVNLRQS ENTSANEKEV EAEFLRLSLG FKCDWFTLEK RVKLEERSRD LAEENLKKEI TNCLKLLESL TPLCEDDNQA QEIIKKLEKS IKFLSQCAAR VASRAEMLGA INQESRVSKA VEVMIQHVEN LKRMYAKEHA ELEELKQVLL QNERSFNPLE

Plays a role in the delivery of peptides to major histocompatibility complex (MHC) class I molecules; this occurs in a transporter associated with antigen processing (TAP)-independent manner. May play a role in taste signal transduction via ITPR3. May play a role during fertilization in pronucleus congression and fusion.

Specifications

Antigen Lrmp
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K52026_16E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene LRMP
Gene Accession No. Q12912
Gene Alias D6Int3; D6Int4; D6Int5; D6Int7; D6Int8; JAW1; LRMP; lymphoid restricted membrane protein; lymphoid-restricted membrane protein; Processed lymphoid-restricted membrane protein; Protein Jaw1
Gene Symbols LRMP
Host Species Mouse
Immunogen Recombinant human IRAG2 (immunogen range is 51-350).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less