Learn More
MilliporeSigma™ Supelco™ LpDNPH S10 Cartridge, Bed wt. 350 mg
Easy to use in the field and laboratory
$313.18 - $1,335.19
Specifications
Basis Weight 350 mg
Volume (Metric) 3 mL
View More Specs
Products 4
Catalog Number Mfr. No. Type Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Packaging Price Quantity & Availability  
111008039
View Documents Promotion Details
MilliporeSigma™ Supelco™
23124U
SPE Cartridge 50/Pk. Bulk Packed
Each for $1,035.56
Only null left
111010101
View Documents Promotion Details
MilliporeSigma™ Supelco™
21026U
SPE Cartridge 10/Pk. Bulk Packed
Each for $313.18
Only null left
111016363
View Documents Promotion Details
MilliporeSigma™ Supelco™
21014
S10 Cartridge 50/Pk. Bulk Packed
Each for $1,335.19
Only null left
111008181
View Documents Promotion Details
MilliporeSigma™ Supelco™
54072U
SPE Cartridge 50/Pk. 1/Zip Bag
Each for $1,325.04
Only null left
Description
Description
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.
Specifications
Specifications
350 mg
3 mL