Learn More
MilliporeSigma™ Supelco™ LpDNPH H Series Cartridges H10, Bed wt. 350 mg
Catalog No. 111007296
Change view
Click to view available options
Volume (Metric):
3 mL
Catalog No. Volume (Metric)
111007296 3 mL
1 options
Catalog No. 111007296 Supplier MilliporeSigma™ Supelco™ Supplier No. 505320U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for sampling carbonyls
The H series of LpDNPH cartridges contain higher loadings of 2,4-DNPH and/or larger bed weights compared the S10 series and Rezorian cartridges. This provides a higher capacity for carbonyls making the H series cartridges the preferred choice for use in high concentration environments. The H series is available in 3 sizes: H10 (350 mg), H30 (1 g) and H300 (10 g) cartridges.
Specifications
Description Bed wt. 350 mg
Quantity 10/Pk.
Volume (Metric) 3 mL