missing translation for 'onlineSavingsMsg'
Learn More

Loxoprofen Sodium Salt Dihydrate 98.0+%, TCI America™

Supplier:  TCI America L025225G

 View more versions of this product

Catalog No. L025225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Loxoprofen Sodium Salt Dihydrate
198°C
C15H21NaO5
25 g
Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate, 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid Sodium Salt
CC(C1=CC=C(C=C1)CC2CCCC2=O)C(=O)[O-].O.O.[Na+]
304.318
CHEBI:31786
≥98.0% (HPLC,T)
226721-96-6
White
MFCD01745788
2811
BAZQYVYVKYOAGO-UHFFFAOYSA-M
sodium;2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate;dihydrate
23674745
268.29
Crystalline Powder

Chemical Identifiers

226721-96-6
304.318
BAZQYVYVKYOAGO-UHFFFAOYSA-M
23674745
sodium;2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate;dihydrate
C15H21NaO5
MFCD01745788
Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate, 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid Sodium Salt
CHEBI:31786
CC(C1=CC=C(C=C1)CC2CCCC2=O)C(=O)[O-].O.O.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CY1678845

TSCA : No

SDS
Documents

Documents

Promotions

Promotions