missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOXL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149819
Description
LOXL3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
LOXL3 is a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.Specifications
| LOXL3 | |
| Polyclonal | |
| Unconjugated | |
| LOXL3 | |
| Lor2; LOW QUALITY PROTEIN: lysyl oxidase homolog 3; LOXL; Loxl2; Loxl3; Lysyl oxidase homolog 3; lysyl oxidase like 3; lysyl oxidase-like 2; lysyl oxidase-like 3; lysyl oxidase-like protein 3; Lysyl oxidase-related protein 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9Z175 | |
| LOXL3 | |
| Recombinant Lysyl Oxidase Like Protein 3 (LOXL3) (His509-Met712). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |