missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOXL2 Recombinant Rabbit Monoclonal Antibody (23GB2610)
Description
LOXL2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | LOXL2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB2610 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | LOXL2 |
| Gene Accession No. | B5DF27, P58022, Q9Y4K0 |
| Gene Alias | 1110004B06Rik; 4930526G11Rik; 9430067E15Rik; LOR2; Loxl2; Lysyl oxidase homolog 2; lysyl oxidase homolog 2; LOW QUALITY PROTEIN: lysyl oxidase homolog 2; lysyl oxidase like 2; lysyl oxidase related 2; lysyl oxidase-like 2; lysyl oxidase-like 2 delta e13; lysyl oxidase-like 2 protein; lysyl oxidase-like protein 2; Lysyl oxidase-related protein 2; lysyl oxidase-related protein WS9-14; WS9-14 |
| Gene Symbols | LOXL2 |
| Show More |