missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems Low Fluorescence Western Membrane (LF PVDF)

Pre-cut LF PVDF membrane. 10 sheets.

Supplier:  Azure Biosystems AC2109

 View more versions of this product

Catalog No. AC2109AZ


Only null left

Description

Description

  • Pre-cut PVDF membranes for Western blotting applications
  • Ideal for chemiluminescent and colorimetric Western blotting. Pre-cut to save you time in the lab. These low-fluorescence PVDF membranes reduce background noise and nonspecific protein binding for improved sensitivity. Available in three different pre-cut sizes.
  • Quantity: 10 membranes per package
Specifications

Chemical Identifiers

Western Blot
PVDF
10 Blots
Sheet
0.45 μm

Specifications

13 cm
Western Blot
PVDF
13 x 18 cm
18 cm
Sheet
0.45 μm
10 Blots
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions