missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Low Density Polyethylene Serum Pipettes
Greiner Bio-One Serum Pipettes are ideally suited for rapid transfer of liquids.
$444.60 - $801.40
Specifications
| Material | LDPE |
|---|---|
| Graduated | Yes |
| Volume (Metric) | 2.5 mL |
| Length (Metric) | 153 mm |
| Catalog Number | Mfr. No. | Sterility | Packaging Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Packaging Type | Price | Quantity | ||||
|
07000569
|
Greiner Bio-One
612301 |
Non-sterile | 500/bag |
Case of 1500 Each for $444.60
|
|
||||
|
07000570
|
Greiner Bio-One
612362 |
Sterile | 25/bag |
Case of 1500 Each for $659.77
|
|
||||
|
07000378
|
Greiner Bio-One
612361 |
Sterile | Individually wrapped |
Case of 800 Each for $801.40
|
|
||||
Description
- With graduation
- Material: Low density polyethylene
- Frapid transfer of liquids
- For single use only
Specifications
| LDPE | |
| Yes | |
| 2.5 mL | |
| 153 mm |