missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Low Density Polyethylene Serum Pipettes

Description
- With graduation
- Material: Low density polyethylene
- Frapid transfer of liquids
- For single use only
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | LDPE |
| Sterility | Sterile |
| For Use With (Application) | Rapid transfer of liquids |
| Graduated | Yes |
| Graduations | 0.5 to 2.5 mL |
| Quantity | 1000/Cs. |
| Packaging Type | 25/bag |
| Color | Translucent |
| Volume (Metric) | 2.5 mL |
| Show More |