Learn More
Invitrogen™ Low Density Lipoprotein from Human Plasma, pHrodo™ Green- and Red-LDL Conjugates
Description
Detect low density lipoprotein (LDL) uptake, endosomal maturation, and trafficking in cells with pHrodo Green-LDL and pHrodo Red-LDL conjugates. These LDL conjugates contain pH-sensitive pHrodo dyes that are weakly fluorescent at neutral pH, yet fluoresce brightly upon exposure to acidic environments, such as those found in endosomes. The pHrodo Green-LDL and Red-LDL conjugates can be used in fluorescence microscopy, flow cytometry, high-throughput screening, and high content analysis experiments.
Features of pHrodo Green-LDL:
• Detectable with GFP/FITC filter set (absorption peak: ∼509 nm, emission peak: ∼533 nm)
• Mutiplexable with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others.
Features of pHrodo Red-LDL:
• Detectable with RFP/TRITC filter set (absorption peak: ∼560 nm, emission peak: ∼585 nm)
• Mutiplexable with a wide variety of blue, green, and far red dyes and reporters such as GFP, Fuo-4, calcein, NucBlue, CellEvent Caspase 3/7 green, Mitosox Green, and Mitotracker Deep Red, among many others.
Both pHrodo Green-LDL and pHrodo Red-LDL can be used in a no-wash protocol, allowing dynamic measurement of LDL uptake and cell trafficking.
Specifications
Specifications
| Content And Storage | • 1 x 200 μL pHrodo Green-LDL, 1 mg/mL Store in refrigerator (2–8°C) and protect from light. |
| Description | Low Density Lipoprotein from Human Plasma, pHrodo Green Conjugate (pHrodo Green-LDL) |
| For Use With (Application) | Cell Analysis |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer, High Content Analysis Instrument |
| Product Type | Cell Labeling Solution |
| Form | Solution |
| Quantity | 200 μL |
| Shipping Condition | Wet Ice |
| Species | Human |
| Conjugate | pHrodo Green |
Frequently Asked Questions (FAQs)
This product does not contain acetylated or oxidized LDL.
pHrodo Red and Green dyes are pH-sensitive dyes. They should only be used on live cells to detect pH of cellular compartments. Upon fixation of the cells, the cell membrane is compromised and you are only measuring the pH of the buffer the samples are held in.
pHrodo Red dye is covalently bound to the LDL.