missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kent Scientific Low Cost Induction Chambers
For use with SomnoSuite Anesthesia System.
Supplier: Kent Scientific SOMNO0720
Includes: Connection tubing
Description
- Sized specifically for mice and rats
- Shatter-resistant plastic
- Easy to clean
- Autoclavable
- Air tight seal
- Connection tubing connects chamber to inlet and outlet of vaporizer
- Resistant to standard anesthesia chemicals
- Small size reduces induction time and waste gas
Applications
Anesthesia, surgery
Warranty
One year warranty including all parts and labor charges
Chemical Identifiers
| Autoclavable | |
| Low Cost Induction Chamber | |
| Connection tubing |
| Anesthesia, Surgery | |
| SomnoSuite Anesthesia System | |
| Shatter-resistant Plastic |
Specifications
| Autoclavable | |
| Anesthesia, Surgery | |
| SomnoSuite Anesthesia System | |
| Shatter-resistant Plastic |
| 2 L | |
| Low Cost Induction Chamber | |
| Connection tubing | |
| 15.8 x 21.6 x 7.6 cm |