missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Losartan Potassium 98.0+%, TCI America™
Supplier: TCI America L023225G
Specifications
| Losartan Potassium | |
| 124750-99-8 | |
| 25 g | |
| MFCD02092704 | |
| GKVBJVGUWMACMS-UHFFFAOYSA-N | |
| [2-butyl-5-chloro-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol;molecular hydrogen;potassium | |
| 131674082 | |
| ≥98.0% (HPLC,T) |
| White-Yellow | |
| Crystalline Powder | |
| C22H25ClKN6O | |
| 2-Butyl-4-chloro-1-[[2′C-(1H-tetrazol-5-yl)-1,1′C-biphenyl-4-yl]methyl]imidazole-5-methanol Potassium Salt | |
| [HH].CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NNN=N4)CO)Cl.[K] | |
| 464.031 | |
| 461.01 |
Chemical Identifiers
| 124750-99-8 | |
| 464.031 | |
| GKVBJVGUWMACMS-UHFFFAOYSA-N | |
| 131674082 | |
| [HH].CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NNN=N4)CO)Cl.[K] |
| C22H25ClKN6O | |
| MFCD02092704 | |
| 2-Butyl-4-chloro-1-[[2′C-(1H-tetrazol-5-yl)-1,1′C-biphenyl-4-yl]methyl]imidazole-5-methanol Potassium Salt | |
| [2-butyl-5-chloro-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol;molecular hydrogen;potassium |
Safety and Handling
RTECSNumber : NI6755100
TSCA : No