missing translation for 'onlineSavingsMsg'
Learn More

Loratadine 98.0+%, TCI America™

Catalog No. L02231G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02231G 1 g
L0223100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02231G Supplier TCI America Supplier No. L02231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79794-75-5
Molecular Formula C22H23ClN2O2
Molecular Weight (g/mol) 382.888
MDL Number MFCD00672869
InChI Key JCCNYMKQOSZNPW-UHFFFAOYSA-N
Synonym loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
PubChem CID 3957
IUPAC Name ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1

Specifications

Melting Point 137°C
Color White
Quantity 1 g
Formula Weight 382.89
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Loratadine
RTECSNumber TM6129200
TSCA No