missing translation for 'onlineSavingsMsg'
Learn More

Loratadine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015561
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015561 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015561 Supplier Merck Emd Millipore Supplier No. PHR13761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Loratadine belongs to the class of H1-antihistamine drugs, with non-sedating properties, and is widely used in the treatment of allergies.

Specifications

CAS 79794-75-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H23N2O2 Cl
MDL Number MFCD00672869
Synonym Loratadine; 4-(8-Chloro-5,6-dihydro-11 H-benzo[5,6 ]cyclohepta[1,2-b]pyridin-11-ylidene-1-piperidinecarboxylic acid ethyl ester; Loratidine)
Type Certified Reference Material
Quantity 1 g
Formula Weight 382.88
Grade Certified Reference Material