missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loratadine, 98+%
Description
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
Specifications
Specifications
| Melting Point | 134°C |
| Color | White |
| Quantity | 1 g |
| Merck Index | 14,5578 |
| Solubility Information | It is soluble in DMSO (50mg/ml),ethanol (77mg/ml at 25°C),water (<1mg/ml at 25°C),chloroform,and methanol. |
| Formula Weight | 382.88 |
| Percent Purity | ≥98% |
| Physical Form | Powder |
| Chemical Name or Material | Loratadine |
Safety and Handling
| RTECSNumber | TM6129200 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only