missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System RNA Cassettes

Catalog No. p-4684348
Encompass
Change view
Click to view available options
Wells:
16 + 1-well
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
BMA57028 16 + 1-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA57028 Supplier Lonza™ Supplier No. 57028
Only null left

Cassettes optimized for RNA. FlashGel™ System RNA Cassettes are recommended for verification and analysis of total RNA, quick checks of native RNA, and checking RNA degradation and mRNA purity.

Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains and hazardous reagents
  • Casettes fully enclose the gel, stain and running buffer
  • Protect samples from contaminating RNases
  • Designed for performance and purity, and are guaranteed RNase-free
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals

Specifications

Concentration 1.2%
Separation Range 0.5 to 9 kb
Wells 16 + 1-well
Formulation 1.2% Agarose
For Use With (Application) Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm
Product Type RNA Cassettes
Show More Show Less