missing translation for 'onlineSavingsMsg'
Learn More

Lomustine 98.0+%, TCI America™

Catalog No. L025150MG
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L025150MG 50 mg
L0251200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L025150MG Supplier TCI America Supplier No. L025150MG
Only null left

Chemical Identifiers

CAS 13010-47-4
Molecular Formula C9H16ClN3O2
Molecular Weight (g/mol) 233.70
MDL Number MFCD00012392
InChI Key GQYIWUVLTXOXAJ-UHFFFAOYSA-N
Synonym 1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea
PubChem CID 3950
ChEBI CHEBI:6520
IUPAC Name 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea
SMILES ClCCN(N=O)C(=O)NC1CCCCC1

Specifications

Melting Point 91°C
Color Yellow
Quantity 50 mg
UN Number 2811
Formula Weight 233.70
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Lomustine
RTECSNumber YS4900000
TSCA No
Recommended Storage Freezer