missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lomerizine Dihydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 180°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 541.46 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lomerizine Dihydrochloride |
Safety and Handling
| RTECSNumber | TK9189000 |
| TSCA | No |
| Recommended Storage | Refrigerator |