Learn More
Brady™ Lockout Tagout Labels: ALERT Signs 5/Package
Catalog No. 19110682
Change view
Click to view available options
Adhesive Type:
Permanent Acrylic, Self-adhesive
Catalog No. Adhesive Type
19110682 Permanent Acrylic, Self-adhesive
1 options
Catalog No. 19110682 Supplier Brady™ Supplier No. 49847
Only null left
Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Languages No Text
Quantity 5/Pk.
Width (Metric) 92.71 mm
Show More Show Less