missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Lockout Hasp (Red)

Catalog No. 19104142
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19104142 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19104142 Supplier Brady™ Supplier No. LH220ARD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hasps allow multiple padlocks to secure a single lockout device.

  • Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
  • Brady™ is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices

Specifications

Product Type Snap-in Hasp
Alternate Part Number NON-CONDUCTIVE LOCKOUT HASPS
Color Red
Material Plastic
Certifications/Compliance OSHA 1910.147
Depth (English) 1 in.
Dimensions (D x W x H) 1 x 2.5 x 4.77 in.
Height (English) 4.77 in.
Quantity 1 Ea.
Width (English) 2.5 in.