missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lobenzarit Disodium Salt 98.0+%, TCI America™
$175.20 - $519.80
Chemical Identifiers
| CAS | 64808-48-6 |
|---|---|
| Molecular Formula | C14H8ClNNa2O4 |
| Molecular Weight (g/mol) | 335.651 |
| MDL Number | MFCD00941422 |
| InChI Key | QPJBONAWFAURGB-UHFFFAOYSA-L |
| Synonym | N-(2-Carboxyphenyl)-4-chloroanthranilic Acid Disodium Salt, Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate |
| PubChem CID | 47462 |
| IUPAC Name | disodium;2-(2-carboxylatoanilino)-4-chlorobenzoate |
| SMILES | C1=CC=C(C(=C1)C(=O)[O-])NC2=C(C=CC(=C2)Cl)C(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L027825MG
|
TCI America
L027825MG |
25 mg |
Each for $175.20
|
|
|||||
|
L0278100MG
|
TCI America
L0278100MG |
100 mg |
Each for $519.80
|
|
|||||
Chemical Identifiers
| 64808-48-6 | |
| 335.651 | |
| QPJBONAWFAURGB-UHFFFAOYSA-L | |
| 47462 | |
| C1=CC=C(C(=C1)C(=O)[O-])NC2=C(C=CC(=C2)Cl)C(=O)[O-].[Na+].[Na+] |
| C14H8ClNNa2O4 | |
| MFCD00941422 | |
| N-(2-Carboxyphenyl)-4-chloroanthranilic Acid Disodium Salt, Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate | |
| disodium;2-(2-carboxylatoanilino)-4-chlorobenzoate |
Specifications
| 64808-48-6 | |
| White-Yellow | |
| MFCD00941422 | |
| N-(2-Carboxyphenyl)-4-chloroanthranilic Acid Disodium Salt, Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate | |
| C1=CC=C(C(=C1)C(=O)[O-])NC2=C(C=CC(=C2)Cl)C(=O)[O-].[Na+].[Na+] | |
| 335.651 | |
| 335.65 | |
| Crystalline Powder |
| 388°C | |
| C14H8ClNNa2O4 | |
| 25 mg | |
| QPJBONAWFAURGB-UHFFFAOYSA-L | |
| disodium;2-(2-carboxylatoanilino)-4-chlorobenzoate | |
| 47462 | |
| ≥98.0% (HPLC,T) | |
| Lobenzarit Disodium Salt |
Safety and Handling
RTECSNumber : DG4975520
TSCA : No