missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIX1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106992 Supplier Invitrogen™ Supplier No. PA5106992
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 93%; Rat: 86%.

LIX1L is a protein coding gene.

Specifications

Antigen LIX1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene LIX1L
Gene Accession No. Q8IVB5
Gene Alias D130027M04Rik; limb and CNS expressed 1 like; Lix1 homolog (chicken) like; Lix1 homolog-like; LIX1L; Lix1-like; LIX1-like protein
Gene Symbols LIX1L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human LIX1L (aa 1-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less