missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ BacLight™ Bacterial Viability Kits

Catalog No. L13152
Change view
Click to view available options
For Use With (Application):
Microscopy
Microscopy, Quantitative Assays
Viability Assay
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
L13152 Viability Assay
L7007 Microscopy
L7012 Microscopy, Quantitative Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L13152 Supplier Invitrogen™ Supplier No. L13152

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LIVE/DEAD™ BacLight™ Bacterial Viability Kits are easy-to-use fluorescent staining kits commonly used for assessing the viability of bacterial cells. These kits provide a two-color fluorescence assay of bacterial viability using green and red fluorescent dyes for use with imaging, flow, and assays.

The LIVE/DEAD BacLight Bacterial Viability Kits are convenient and easy-to-use kits for monitoring the viability of bacterial populations based on the membrane integrity of the cell. These kits provide a two-color fluorescence assay of bacterial viability for a diverse array of bacterial genera. Cells with a compromised membrane that are dead or dying will stain red with propidium iodide, whereas cells with an intact membrane will stain green with SYTO™ 9. These kits enable researchers to differentiate between live and dead bacteria within minutes, even in mixed populations with diverse bacterial types through fluorescent imaging, flow cytometry, and microplate assays.

The LIVE/DEAD BacLight Bacterial Viability Kits utilize mixtures of our SYTO™ 9 green-fluorescent nucleic acid stain and the red-fluorescent nucleic acid stain, propidium iodide. These stains differ both in their spectral characteristics and in their ability to penetrate healthy bacterial cells. When used alone, the SYTO 9 stain generally labels all bacteria in a population — those with intact membranes and those with damaged membranes. In contrast, propidium iodide penetrates only bacteria with damaged membranes, causing a reduction in the SYTO 9 stain fluorescence when both dyes are present.

The LIVE/DEAD Bac Light Bacterial Viability Kits have been thoroughly tested with a variety of organisms and under several conditions. The kits are ideal for fluorescence microscopy as the live (green fluorescent) and dead (red fluorescent) cells may be viewed separately. They are also suitable for quantitative analysis with a fluorometer, fluorescence microplate reader, flow cytometer, or other instrumentation.

Traditional methods of assessing bacterial viability rely on metabolic characteristics or membrane integrity. However, due to differences in morphology, cytology, and physiology among the many bacterial genera, developing a universally applicable direct-count viability assay has been challenging. The LIVE/DEAD BacLight Bacterial Viability Kits based on membrane integrity allow researchers to distinguish live and dead bacteria easily, reliably, and quantitatively in minutes, even in a mixed population with diverse bacterial types.

The LIVE/DEAD BacLight Bacterial Viability Kit is available in three different formats:

LIVE/DEAD BacLight Bacterial Viability Kit, for microscopy (Cat. No. L7007)
The LIVE/DEAD BacLight Bacterial Viability Kit, for microscopy includes two different premixed solutions of the SYTO 9 and propidium iodide (PI) in 300 μL of DMSO. The fluorescent dyes are mixed at different proportions in each one: Component A – 1.67 mM SYTO 9 & 1.67 mM PI, Component B – 1.67 mM STYO 9 & 18.3 mM PI. This kit is available for customers who have already developed protocols using the premixed dye formulation.

LIVE/DEAD BacLight Bacterial Viability Kit, for microscopy & quantitative assays (Cat. No. L7012)
The LIVE/DEAD BacLight Bacterial Viability Kit, for microscopy & quantitative assays includes two separate solutions of the SYTO 9 (3.34 mM) and propidium iodide (20 mM) dyes in 300 μL of DMSO. The separate staining components enables flexibility for optimization and facilitates the calibration of bacterial fluorescence for quantitative procedures.

LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L13152)
For added convenience, the LIVE/DEAD BacLight Bacterial Viability Kit contains 10 pairs of the premeasured separate SYTO 9 and propidium iodide dyes in sealed polyethylene transfer pipets. In addition to the convenient packaging in the sealed pipets, the fluorescent dyes are supplied dry, without DMSO or other solvents. This formulation does not require refrigeration and remains stable at temperatures ≤–20°C once reconstituted.

All three kits come with 10mL of BacLight mounting oil for bacteria immobilized on membranes. The refractive index at 25°C is 1.517 ± 0.003.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

At the recommended reagent dilutions and volumes, kits L7007 and L7012 contain sufficient material to perform ≥1000 individual tests in 96-well assay plates, many more tests by fluorescence microscopy or ~200 tests by flow cytometry.

In kit L13152, each applicator pair contains sufficient dye to perform 50 individual tests in a 96-well assay plate, ~1000 assays by fluorescence microscopy or 10 tests by flow cytometry.

The LIVE/DEAD BacLight Bacterial Viability Kits have been tested on the following bacterial species: Bacillus cereus, B. subtilis, Clostridium perfringens, Escherichia coli, Klebsiella pneumoniae, Micrococcus luteus, Mycobacterium phlei, Pseudomonas aeruginosa, P. syringae, Salmonella oranienburg, Serratia marcescens, Shigella sonnei, Staphylococcus aureus and Streptococcus pyogenes. All these bacterial types have shown a good correlation between the results obtained with the LIVE/DEAD BacLight Bacterial Viability Kits and those obtained with standard plate counts.

These tests were performed on logarithmically growing cultures of organisms. In addition, we have received favorable reports from researchers who have used these kits with: Agrobacterium tumefaciens, Edwardsiella ictaluri, Eurioplasma eurilytica, Lactobacillus sp., Mycoplasma hominus, Propionibacterium sp., Proteus mirabilis and Zymomonas sp.

Order Info

Shipping Condition: Room temperature. This stain formulation does not require refrigeration and is chemically stable even in poor conditions.

Specifications

Cell Type Bacteria
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Fluorescence Microplate Reader, Flow Cytometer
Product Type Bacterial Viability Kit
Dye Type Other Label(s) or Dye(s)
Format Tube(s)
Product Line BacLight, LIVE/DEAD
Quantity 10 Applicator Sets kit
Shipping Condition Room Temperature
Show More Show Less
When using the LIVE/DEAD BacLight kit for bacterial viability, with SYTO 9 and propidium iodide (PI), why are some cells detected with both dyes? Shouldn't live cells be green and dead cells be red?

SYTO 9 dye will enter all cells, live or dead. PI only enters cells with compromised membranes (dead cells or damaged cells). First, perform single color staining and examine under both filter sets. Longpass filters or the use of too much dye may result in excessive bleedthrough of the green dye emission into the red channel and the emission of PI in the green channel. Use narrower bandpass filters if possible, and use lower concentrations of the dye. Some live cells may take up PI by engulfment; avoid extended incubation times with the dye. Apply PI to a live cell culture and optimize incubation times to limit engulfment.

I want to use propidium iodide and SYTO 9 to do LIVE/DEAD testing of a bacterial sample. Will anaerobic conditions adversely affect the assay?

Oxygen content should not affect the binding of propidium iodide and SYTO 9 to nucleic acids. SYTO 9 will label all cells, and propidium iodide will label only dead cells or cells with a compromised membrane.

I have a LIVE/DEAD BacLight Bacterial Viability kit that has SYTO 9 and propidium iodide in it. Will I be able to stain eukaryotic cells that have engulfed bacteria and determine if the bacteria are alive or dead using this kit?

Unfortunately, no. SYTO 9 will label the nuclei of live or dead cells, including the eukaryotic cells. Propidium iodide is cell impermeant, and will only enter dead cells. If the eukaryotic cells are dead, they will label with propidium iodide as well. If the eukaryotic cells are alive, propidium iodide will not be able to enter and thus will not label the bacteria inside, whether the bacteria are alive or dead. We are not aware of any way to do a viability assay of bacteria once they have been engulfed by cells.

How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

What bacterial parameters can I look at by flow cytometry?

You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).

Can I look at bacteria and yeast on a flow cytometer?

Yes, you can. We offer:

-Counting assays: Bacteria Counting Kit, for flow cytometry (Cat. No. B7277) or LIVE/DEAD BacLight Bacterial Viability and Counting Kit, for flow cytometry (Cat. No. L34856).
-Viability/vitality assays: LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L13152).
-Membrane potential: BacLight Bacterial Membrane Potential Kit (Cat. No. B34950).
-Yeast viability/vitality assays: LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry (Cat. No. L34952), FungaLight Yeast CFDA, AM/Propidium Iodide Vitality Kit (Cat. No. F34953)


For Research Use Only. Not for use in diagnostic procedures.