missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation

Catalog No. L34976
Change view
Click to view available options
Quantity:
200 Assays
400 Assays
80 assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L34976 400 Assays
501121531 80 assays
L10119 200 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L34976 Supplier Invitrogen™ Supplier No. L34976

Determines the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation.

The LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation. This kit has been optimized and validated for use with a red laser flow cytometer.

  • Stable—dyes are freeze dried in separate vials to maintain stability
  • Robust—staining pattern is the same before and after fixation
  • Bright signal—allows for easy distinction between live/dead cells in single channel

Stable

Unlike products that are sold in solution, the LIVE/DEAD™ Fixable Near-IR Stain has been conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time. Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated. If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles.

Robust

Dead cell discriminator stains can lose sensitivity after treatment with fixatives such as formaldehyde or ethanol-based fixation methods required for intracellular phosphorylation studies. The LIVE/DEAD™ Fixable Near-IR Stain is an amine reactive dye that binds covalently to intracellular and extracellular amines, and the staining pattern is preserved following formaldehyde fixation.

Optimal brightness

The LIVE/DEAD™ Fixable Near-IR stain was selected based on its fluorescent properties to provide a bright signal when excited with a red laser. The near-IR fluorescent reactive dye has an excitation maximum of ∼633 nm making it ideal for use with the red or HeNe laser with an emission of ∼775 nm. Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments.

How it works

In cells with compromised membranes, the dye reacts with free amines both in the cell interior and on the cell surface, yielding intense fluorescent staining. In viable cells, the dye's reactivity is restricted to the cell-surface amines, resulting in less intense fluorescence. The difference in intensity is typically greater than 50-fold between live and dead cells, allowing for easy discrimination.

Colors available

LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet your multi-color panel needs.

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Near-IR
Content And Storage Contains 10 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Description LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Stain
Dye Type LIVE/DEAD™ FixableNear-IR Dead Cell Stain
Emission 775 nm
Excitation Wavelength Range 633 nm
Form Solid
Format Tube(s)
Product Line LIVE/DEAD
Quantity 400 Assays
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less
How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

Which cell viability kits are compatible with fixation?

The LIVE/DEAD Fixable kits for flow cytometry analysis are compatible with fixation. These kits use amine-reactive cell-impermeant dyes that stain the cell surface of live cells and also the cytosol of dead cells-live cells are dim and dead cells are bright. Since the dye is covalently bound to the cells, it will be retained after fixation. Unfortunately, this method does not work well for imaging-based assays, as all cells are stained and it is difficult to distinguish bright dead cells from dim live cells with a microscope. Ethidium monoazide (EMA; Cat No. E1374) is a cell impermeant nucleic acid stain that can be applied to live cultures and stains only dead cells. After incubation and washing away unbound dye, the cells can be exposed to light to photoactivate EMA to crosslink to dead cell DNA. After crosslinking to dead cell DNA, the samples may be fixed and permeabilized. Image-IT DEAD Green Viability Stain (Cat. No. I10291) for imaging and high-content screening (HCS) analysis is a live-cell impermeant DNA binding dye that is compatible with fixation and permeabilization with good retention up to 48 hours. We also have a LIVE/DEAD Reduced Biohazard Cell Viability Kit (Cat. No. L7013) for imaging and flow analysis that contains two DNA binding dyes, SYTO 10 and Dead Red, that are sufficiently retained to be analyzed soon after 4% glutaraldehyde fixation.
Note: In general, DNA-binding dyes and calcein AM are not compatible with fixation, as these dyes are not covalently bound to components of the cell and will thus slowly diffuse out of cells after fixation, gradually staining all cells as dead.

For Research Use Only. Not for use in diagnostic procedures.