Learn More
Invitrogen™ LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation
Description
The LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation. This kit has been optimized and validated for use with a red laser flow cytometer.
- Stable—dyes are freeze dried in separate vials to maintain stability
- Robust—staining pattern is the same before and after fixation
- Bright signal—allows for easy distinction between live/dead cells in single channel
Stable
The product is sold in tubes with DMSO and in lyophilized flow tubes and plates which require no DMSO. Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use material once rehydrated.
Robust
Dead cell discriminator stains can lose sensitivity after treatment with fixatives such as formaldehyde or ethanol-based fixation methods required for intracellular phosphorylation studies. The LIVE/DEAD™ Fixable Near-IR Stain is an amine reactive dye that binds covalently to intracellular and extracellular amines, and the staining pattern is preserved following formaldehyde fixation.
Optimal brightness
The LIVE/DEAD™ Fixable Near-IR stain was selected based on its fluorescent properties to provide a bright signal when excited with a red laser. The near-IR fluorescent reactive dye has an excitation maximum of ∼633 nm making it ideal for use with the red or HeNe laser with an emission of ∼775 nm. Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments.
How it works
In cells with compromised membranes, the dye reacts with free amines both in the cell interior and on the cell surface, yielding intense fluorescent staining. In viable cells, the dye's reactivity is restricted to the cell-surface amines, resulting in less intense fluorescence. The difference in intensity is typically greater than 50-fold between live and dead cells, allowing for easy discrimination.
Colors available
LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet your multi-color panel needs.
Specifications
Specifications
| Cell Permeability | Impermeant |
| Cell Type | Eukaryotic Cells |
| Color | Near-IR |
| Content And Storage | Contains 5 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO. Store at -20°C. |
| Description | LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit, for 633 or 635 nm excitation |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Stain |
| Dye Type | LIVE/DEAD™ FixableNear-IR Dead Cell Stain |
| Show More |
Frequently Asked Questions (FAQs)
Heat killing is commonly used. Place your cells in a tube in buffer and heat at 60oC for 20 minutes. You can also kill your cells by fixing them with ice cold 70% ethanol for 15 minutes. The ethanol-killed cells can then be stored at -20oC until needed, at which point you wash out the ethanol and replace with buffer.
This dye gives a dim surface label for live cells, but is internalized and gives a brighter signal for dead cells. Flow cytometry is a very sensitive technique and can easily distinguish between the two populations. Microscopy is not as sensitive and may not be able to distinguish the cells because of a less sensitive detector.
There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.
Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.
The LIVE/DEAD Fixable kits for flow cytometry analysis are compatible with fixation. These kits use amine-reactive cell-impermeant dyes that stain the cell surface of live cells and also the cytosol of dead cells-live cells are dim and dead cells are bright. Since the dye is covalently bound to the cells, it will be retained after fixation. Unfortunately, this method does not work well for imaging-based assays, as all cells are stained and it is difficult to distinguish bright dead cells from dim live cells with a microscope. Ethidium monoazide (EMA; Cat No. E1374) is a cell impermeant nucleic acid stain that can be applied to live cultures and stains only dead cells. After incubation and washing away unbound dye, the cells can be exposed to light to photoactivate EMA to crosslink to dead cell DNA. After crosslinking to dead cell DNA, the samples may be fixed and permeabilized.
Image-IT DEAD Green Viability Stain (Cat. No. I10291) for imaging and high-content screening (HCS) analysis is a live-cell impermeant DNA binding dye that is compatible with fixation and permeabilization with good retention up to 48 hours.
We also have a LIVE/DEAD Reduced Biohazard Cell Viability Kit (Cat. No. L7013) for imaging and flow analysis that contains two DNA binding dyes, SYTO 10 and Dead Red, that are sufficiently retained to be analyzed soon after 4% glutaraldehyde fixation.
Note: In general, DNA-binding dyes and calcein AM are not compatible with fixation, as these dyes are not covalently bound to components of the cell and will thus slowly diffuse out of cells after fixation, gradually staining all cells as dead.
Many antibodies and stains will label dead cells. This will give you misleading data if you do not exclude the dead cells from your analysis. Of course, if you are labeling fixed cells, they are already dead and you do not need a viability stain. However, if you label your cells prior to fixation, then you need to use one of the LIVE/DEAD Fixable Dead Cell Stains.
For Research Use Only. Not for use in diagnostic procedures.