missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation DFS Item

The LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

Supplier:  Molecular Probes™ L34965

 View more versions of this product

Catalog No. 501121525

Only null left

Description

Description

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a violet laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Low compensation

  • The LIVE/DEAD™ Fixable Aqua Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser
  • The aqua-fluorescent reactive dye has an excitation maximum of ∼375 nm, but it is excited well with the 405 nm violet laser
  • It has an emission maxima of ∼512 nm, so it can be collected in the second channel on most violet laser flow cytometers
Specifications

Specifications

Impermeant
Aqua
LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation
Viability Assay
Stain
526
Solid
LIVE/DEAD
Room Temperature
Eukaryotic Cells
Contains 2 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Fluorescence
Flow Cytometer
LIVE/DEAD™ Fixable Aqua Dead Cell Stain
375 nm
Tube
80 Assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.