missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium zirconium oxide, 94% min, Thermo Scientific Chemicals
Description
Lithium zirconium oxide is used in the separation and absorption of carbon dioxide. It is used in the manufacture of dielectric bodies and insulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Cream |
| Physical Form | Powder |
| Quantity | 25 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 153.1 |
| Percent Purity | ≤94% |
| Odor | Odorless |
| Chemical Name or Material | Lithium zirconium oxide |
Safety and Handling
| EINECSNumber | 234-760-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only