accessibility menu, dialog, popup

UserName

LiLiOZrOO

CAS RN 12031-83-3

IUPAC Name: dilithium(1+) oxozirconiumbis(olate)
Synonyms: dilithium(1+) oxozirconiumbis(olate)
Molecular Weight (g/mol): 153.1
Molecular Formula: Li2O3Zr
InChi Key: PSKLXRHTVCSWPJ-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[O-][Zr]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results