missing translation for 'onlineSavingsMsg'
Learn More

Lithium zirconium oxide, 94% min, Thermo Scientific Chemicals

Catalog No. AA3937714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3937714 25 g
AA3937722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3937714 Supplier Thermo Scientific Chemicals Supplier No. 03937714
Only null left

Lithium zirconium oxide is used in the separation and absorption of carbon dioxide. It is used in the manufacture of dielectric bodies and insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12031-83-3
Molecular Formula Li2O3Zr
Molecular Weight (g/mol) 153.10
MDL Number MFCD00016182
InChI Key PSKLXRHTVCSWPJ-UHFFFAOYSA-N
Synonym lithium zirconate, dilithium dioxido oxo zirconium, 2li.o3zr, lithium zirconium oxide, dilithotab oxozirconiumbis olate, lithium zirconium oxide min, dilithium 1+ ion oxozirconiumbis olate, dilithium 1+ oxozirconiumbis olate
PubChem CID 6093659
IUPAC Name dilithium;dioxido(oxo)zirconium
SMILES [Li+].[Li+].[O-][Zr]([O-])=O

Specifications

Color White to Cream
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 153.1
Percent Purity ≤94%
Odor Odorless
Chemical Name or Material Lithium zirconium oxide
EINECSNumber 234-760-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only